Anti-RAB30 Mouse Monoclonal Antibody [clone: 3B1]

Lieferant: OriGene
ORIGCF505347EA 749 EUR
ORIGCF505347
Anti-RAB30 Mouse Monoclonal Antibody [clone: 3B1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RAB30
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR