Anti-NEK9 Mouse Monoclonal Antibody [clone: 7G7]

Lieferant: OriGene
ORIGCF503367EA 749 EUR
ORIGCF503367
Anti-NEK9 Mouse Monoclonal Antibody [clone: 7G7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NEK9
Clonality: Monoclonal
Clone: 7G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR