Anti-ATG3 Mouse Monoclonal Antibody [clone: 3H1]

Lieferant: OriGene
ORIGCF503383EA 749 EUR
ORIGCF503383
Anti-ATG3 Mouse Monoclonal Antibody [clone: 3H1]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATG3
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR