Anti-ANAPC2 Mouse Monoclonal Antibody [clone: 1B11]

Lieferant: OriGene
ORIGCF503406EA 770 EUR
ORIGCF503406
Anti-ANAPC2 Mouse Monoclonal Antibody [clone: 1B11]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ANAPC2
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR