Anti-PLDN Mouse Monoclonal Antibody [clone: 1E2]

Lieferant: OriGene
ORIGCF504156EA 749 EUR
ORIGCF504156
Anti-PLDN Mouse Monoclonal Antibody [clone: 1E2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PLDN
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR