Anti-IGJ Mouse Monoclonal Antibody [clone: 3C7]
Lieferant: OriGene
ORIGCF504168EA 749 EUR
ORIGCF504168
Anti-IGJ Mouse Monoclonal Antibody [clone: 3C7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: IGJ
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human