Anti-LIMK1 Mouse Monoclonal Antibody [clone: 8C8]

Lieferant: OriGene
ORIGCF502978EA 749 EUR
ORIGCF502978
Anti-LIMK1 Mouse Monoclonal Antibody [clone: 8C8]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LIMK1
Clonality: Monoclonal
Clone: 8C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR