Anti-CCM2 Mouse Monoclonal Antibody [clone: 1E9]

Lieferant: OriGene
ORIGCF503417EA 749 EUR
ORIGCF503417
Anti-CCM2 Mouse Monoclonal Antibody [clone: 1E9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CCM2
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR