Anti-PDE4B Mouse Monoclonal Antibody [clone: 2B10]

Lieferant: OriGene
ORIGCF503446EA 749 EUR
ORIGCF503446
Anti-PDE4B Mouse Monoclonal Antibody [clone: 2B10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4B
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR