Anti-SSX5 Mouse Monoclonal Antibody [clone: 1D6]

Lieferant: OriGene
ORIGCF504221EA 770 EUR
ORIGCF504221
Anti-SSX5 Mouse Monoclonal Antibody [clone: 1D6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SSX5
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR