Anti-SOCS3 Mouse Monoclonal Antibody [clone: 7E10]
Lieferant: OriGene
ORIGCF503057EA 749 EUR
ORIGCF503057
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 7E10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 7E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human