Anti-KIF25 Mouse Monoclonal Antibody [clone: 2C9]

Lieferant: OriGene
ORIGCF505426EA 770 EUR
ORIGCF505426
Anti-KIF25 Mouse Monoclonal Antibody [clone: 2C9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: KIF25
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR