Anti-CYP2J2 Mouse Monoclonal Antibody [clone: 3A3]

Lieferant: OriGene
ORIGCF503483EA 749 EUR
ORIGCF503483
Anti-CYP2J2 Mouse Monoclonal Antibody [clone: 3A3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP2J2
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR