Anti-IDS Mouse Monoclonal Antibody [clone: 1D9]

Lieferant: OriGene
ORIGCF504257EA 749 EUR
ORIGCF504257
Anti-IDS Mouse Monoclonal Antibody [clone: 1D9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IDS
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR