Anti-PECI Mouse Monoclonal Antibody [clone: 1C8]

Lieferant: OriGene
ORIGCF504275EA 749 EUR
ORIGCF504275
Anti-PECI Mouse Monoclonal Antibody [clone: 1C8]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PECI
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR