Anti-RUVBL2 Mouse Monoclonal Antibody [clone: 1A6]

Lieferant: OriGene
ORIGCF504281EA 749 EUR
ORIGCF504281
Anti-RUVBL2 Mouse Monoclonal Antibody [clone: 1A6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RUVBL2
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR