Anti-GSPT2 Mouse Monoclonal Antibody [clone: 2G9]
Lieferant: OriGene
ORIGCF503066EA 764 EUR
ORIGCF503066
Anti-GSPT2 Mouse Monoclonal Antibody [clone: 2G9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: GSPT2
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human