Anti-LSM1 Mouse Monoclonal Antibody [clone: 1A9]

Lieferant: OriGene
ORIGCF503107EA 749 EUR
ORIGCF503107
Anti-LSM1 Mouse Monoclonal Antibody [clone: 1A9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LSM1
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR