Anti-PSMA2 Mouse Monoclonal Antibody [clone: 3B3]

Lieferant: OriGene
ORIGCF505471EA 749 EUR
ORIGCF505471
Anti-PSMA2 Mouse Monoclonal Antibody [clone: 3B3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: PSMA2
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR