Anti-GADD45G Mouse Monoclonal Antibody [clone: 1F2]
Lieferant: OriGene
ORIGCF505482EA 749 EUR
ORIGCF505482
Anti-GADD45G Mouse Monoclonal Antibody [clone: 1F2]
Antikörper
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: GADD45G
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human