Anti-MTFMT Mouse Monoclonal Antibody [clone: 1B10]

Lieferant: OriGene
ORIGCF503546EA 749 EUR
ORIGCF503546
Anti-MTFMT Mouse Monoclonal Antibody [clone: 1B10]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: MTFMT
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR