Anti-UNG Mouse Monoclonal Antibody [clone: 2F3]

Lieferant: OriGene
ORIGCF503567EA 749 EUR
ORIGCF503567
Anti-UNG Mouse Monoclonal Antibody [clone: 2F3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: UNG
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR