Anti-NLN Mouse Monoclonal Antibody [clone: 3H4]

Lieferant: OriGene
ORIGCF504327EA 749 EUR
ORIGCF504327
Anti-NLN Mouse Monoclonal Antibody [clone: 3H4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NLN
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR