Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: 4G4]

Lieferant: OriGene
ORIGCF504339EA 770 EUR
ORIGCF504339
Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: 4G4]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TMPRSS5
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR