Anti-BTLA Mouse Monoclonal Antibody [clone: 4D6]

Lieferant: OriGene
ORIGCF505536EA 749 EUR
ORIGCF505536
Anti-BTLA Mouse Monoclonal Antibody [clone: 4D6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: BTLA
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR