Anti-GADD45G Mouse Monoclonal Antibody [clone: 7F3]

Lieferant: OriGene
ORIGCF505546EA 749 EUR
ORIGCF505546
Anti-GADD45G Mouse Monoclonal Antibody [clone: 7F3]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: GADD45G
Clonality: Monoclonal
Clone: 7F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR