Anti-MTFMT Mouse Monoclonal Antibody [clone: 2A2]

Lieferant: OriGene
ORIGCF503571EA 749 EUR
ORIGCF503571
Anti-MTFMT Mouse Monoclonal Antibody [clone: 2A2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MTFMT
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR