Anti-PTK2 Mouse Monoclonal Antibody [clone: 4G7]

Lieferant: OriGene
ORIGCF506182EA 764 EUR
ORIGCF506182
Anti-PTK2 Mouse Monoclonal Antibody [clone: 4G7]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PTK2
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR