Anti-LYZ Mouse Monoclonal Antibody [clone: 2C2]

Lieferant: OriGene
ORIGCF506197EA 764 EUR
ORIGCF506197
Anti-LYZ Mouse Monoclonal Antibody [clone: 2C2]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: LYZ
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR