Anti-NCAM1 Mouse Monoclonal Antibody [clone: 1G4]

Lieferant: OriGene
ORIGCF506208EA 764 EUR
ORIGCF506208
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 1G4]
Antikörper
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 1G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR