Anti-EGFR Mouse Monoclonal Antibody [clone: 2G8]

Lieferant: OriGene
ORIGCF506209EA 764 EUR
ORIGCF506209
Anti-EGFR Mouse Monoclonal Antibody [clone: 2G8]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR