Anti-KRT20 Mouse Monoclonal Antibody [clone: 5E6]

Lieferant: OriGene
ORIGCF802655EA 764 EUR
ORIGCF802655
Anti-KRT20 Mouse Monoclonal Antibody [clone: 5E6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: KRT20
Clonality: Monoclonal
Clone: 5E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR