Anti-PGR Mouse Monoclonal Antibody [clone: 4B3]

Lieferant: OriGene
ORIGCF802702EA 764 EUR
ORIGCF802702
Anti-PGR Mouse Monoclonal Antibody [clone: 4B3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:200

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 4B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR