Anti-SCAMP2 Mouse Monoclonal Antibody [clone: 1H7]

Lieferant: OriGene
ORIGCF506990EA 786 EUR
ORIGCF506990
Anti-SCAMP2 Mouse Monoclonal Antibody [clone: 1H7]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: SCAMP2
Clonality: Monoclonal
Clone: 1H7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR