Anti-CHAC1 Mouse Monoclonal Antibody [clone: 1G2]

Lieferant: OriGene
ORIGCF507009EA 786 EUR
ORIGCF507009
Anti-CHAC1 Mouse Monoclonal Antibody [clone: 1G2]
Antikörper
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CHAC1
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR