Anti-CLDN6 Mouse Monoclonal Antibody [clone: 3G3]

Lieferant: OriGene
ORIGCF507012EA 764 EUR
ORIGCF507012
Anti-CLDN6 Mouse Monoclonal Antibody [clone: 3G3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: CLDN6
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR