Anti-HERPUD1 Mouse Monoclonal Antibody [clone: 1A2]
Lieferant: OriGene
ORIGCF507019EA 764 EUR
ORIGCF507019
Anti-HERPUD1 Mouse Monoclonal Antibody [clone: 1A2]
Antikörper
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: HERPUD1
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat