Anti-CEP68 Mouse Monoclonal Antibody [clone: 5B4]
Lieferant: OriGene
ORIGCF507021EA 786 EUR
ORIGCF507021
Anti-CEP68 Mouse Monoclonal Antibody [clone: 5B4]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100
Type: Primary
Antigen: CEP68
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human