Anti-JUN Mouse Monoclonal Antibody [clone: 3B3]

Lieferant: OriGene
ORIGCF800576EA 764 EUR
ORIGCF800576
Anti-JUN Mouse Monoclonal Antibody [clone: 3B3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR