Anti-JUN Mouse Monoclonal Antibody [clone: 7B10]

Lieferant: OriGene
ORIGCF800585EA 764 EUR
ORIGCF800585
Anti-JUN Mouse Monoclonal Antibody [clone: 7B10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 7B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR