Anti-JUN Mouse Monoclonal Antibody [clone: 5E9]
Lieferant: OriGene
ORIGCF800591EA 764 EUR
ORIGCF800591
Anti-JUN Mouse Monoclonal Antibody [clone: 5E9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 5E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human