Anti-JUN Mouse Monoclonal Antibody [clone: 9H10]
Lieferant: OriGene
ORIGCF800593EA 764 EUR
ORIGCF800593
Anti-JUN Mouse Monoclonal Antibody [clone: 9H10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500
Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 9H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human