Anti-HBS1L Mouse Monoclonal Antibody [clone: 1G7]

Lieferant: OriGene
ORIGCF800608EA 764 EUR
ORIGCF800608
Anti-HBS1L Mouse Monoclonal Antibody [clone: 1G7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HBS1L
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR