Anti-SF3A1 Mouse Monoclonal Antibody [clone: 3F4]

Lieferant: OriGene
ORIGCF800622EA 764 EUR
ORIGCF800622
Anti-SF3A1 Mouse Monoclonal Antibody [clone: 3F4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: SF3A1
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR