Anti-CD33 Mouse Monoclonal Antibody [clone: 2E4]

Lieferant: OriGene
ORIGCF506329EA 764 EUR
ORIGCF506329
Anti-CD33 Mouse Monoclonal Antibody [clone: 2E4]
Antikörper
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR