Anti-ACY1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA323595EA 564 EUR
ORIGTA323595
Anti-ACY1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:200-1000Immunochemistry: 1:25-100

Type: Primary
Antigen: ACY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR