Anti-CDKN1A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA325348EA 564 EUR
ORIGTA325348
Anti-CDKN1A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: CDKN1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR