Anti-SYK Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA323860EA 543 EUR
ORIGTA323860
Anti-SYK Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:100-200

Type: Primary
Antigen: SYK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR