Anti-HSF2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA323863EA 564 EUR
ORIGTA323863
Anti-HSF2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:500-2000

Type: Primary
Antigen: HSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR