Anti-RPS27A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA323865EA 564 EUR
ORIGTA323865
Anti-RPS27A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: RPS27A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR